Learn More
Buehler IsoCut Fluid, 32oz [0.95L]
Supplier: Buehler 111193032
This item has a minimum qty of 7 per supplier requirements.
IsoCut Fluid is an oil based coolant only for use with the IsoMet LS or any saw with a maximum speed of 500rpm. 32 oz.