Description
- Specifically calibrated for use with LINK 7
- Extended capacity
- 10 year warranty contingent upon a 2 year service cycle
- Larger capacity for liquid anesthetic means filling the vaporizer less often which, in turn, reduces exposure to anesthetic gases.
- Available with either funnel fill or pin index/key fill systems.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| For Use With (Equipment) | TECH 3 and Link 7 |
| Size | 250 mL |
| Model | Pin index/key fill |