Learn More
Thomas Scientific IsoFreeze Microcentrifuge Tube Rack, Pink
SDP
Supplier:  Thomas Scientific 1148D59
Encompass
Maintains a sample temperature of less than 4ºC for almost 4 hours (with lid on rack) when used at room temperature. Color-change materia enables you to see how cold your samples are at a glance. Rack comes with lid.
Catalog No. NC1895998
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.