Learn More
Poltex Isolation Gown Dispenser
SDP
Catalog No. 21377560
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
21-377-560 Wall Mount
21-377-558 Magnetic Mount
21-377-559 VHB Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-377-560 Supplier Poltex Supplier No. GOWN1W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Poltex Gown Dispenser holds a stack of isolation gowns or coveralls.
Hold a stack of isolation gowns or coveralls.
Durable and cleanable PETG construction.
Choose mounting method:
  • Wall mount (screws/anchors included)
  • Magnets (powerful magnets indcluded)
  • VHB Tape (very high bond tape included)
Specifications
Product Type Bulk Dispenser
Color Clear
Holds Isolation Gowns or Coveralls
Material PETG
Mounting Wall Mount
No. of Compartments 1
Product Style Open Top and Front
Sterility Non-sterile