Learn More
Description
Benefits:
- Blood culture system for the collection and isolation of microorganisms from the blood. The ISOLATOR™ pediatric tube is used to concentrate and deliver microbial organisms to conventional growth media for purposes of isolation, identification and susceptibility testing.
- When compared to conventional broth systems, lysis centrifugation can reduce the time to isolation by 24 hours
Specifications
Specifications
| Product Type | Microbial Tube |
| Quantity | 25 Pk. |
| Storage Requirements | 2°C to 40°C |
| Tube Size | Pediatric |
| Volume (Metric) | 1.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.