Learn More
TARGETMOL CHEMICALS INC ISOLIENSININE 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S1103
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Isoliensinine (Isoliensinin) a natural phenolic bisbenzyltetrahydroisoquinoline alkaloid has received considerable attention for its potential biological effects such as antioxidant and anti-HIV activities. 2. Isoliensinine possesses an anti-proliferative effect which is related to the decrease of the overexpression of growth factors PDGF-beta bFGF proto-oncogene c-fos c-myc and hsp7. purity: 98%
Catalog No. 50-257-6641
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.