Learn More
Sigma Aldrich Fine Chemicals Biosciences Isomalt, 64519-82-0, MFCD00190708, 3G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences I0465000
Encompass
Molecular Formula: C12H24O11, Molecular Weight: 344.31, Format: Neat, Storage Temperature: 2-8°C
Catalog No. NC0274511
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.