Learn More
TARGETMOL CHEMICALS INC ISOMAZOLE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T276325MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Isomazole is a novel orally available phosphodiesterase (PDE) inhibitor with calcium-sensitizing properties that inhibits PDE3 and PDE4. purity: 99%
Catalog No. 50-258-0592
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.