Learn More
Glass Expansion IsoMist-MS Agilent 7850/7900/8900 ICP-MS
SDP
Supplier:  Glass Expansion KT1201
Encompass_Preferred
IsoMist-MS for Agilent 7850/7900/8900 ICP-MS. The IsoMist-MS™ is a compact and lightweight spray chamber design that has improved transport and washout efficiency for ICP-MS. Having interchangeable Tracey™ DC Spray Chambers available in Glass, PEEK, PFA or Quartz, the IsoMist-MS can handle any ICP-MS application. Compatible with the Agilent 7850/7900/8900, and operates by simply using your exisiting software settings.The novel design (Patent Pending) Jet Vortex Interface (JVI™), adds a highly efficient Aerosol Filtration accessory to the IsoMist-MS, working in conjunction with the existing auxiliary gas options of the Agilent ICP-MS. The JVI accessory allows the analyst to optimally fine-tune their instrument parameters, easily achieving oxide ratios well below 1% and minimizing deposition on the interface cones. A must have accessory for analyzing “real world” high-matrix samples.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2967
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.