Learn More
Glass Expansion IsoMist XS Assembly (LOL)
SDP
Supplier:  Glass Expansion 208094951
Encompass_Preferred
IsoMist XS Assembly (LOL). The IsoMist XS assembly is programmable and temperature-controlled, with a choice of cyclonic spray chamber (Glass, Quartz, or PFA, not included) now featuring an improved thermodynamic design that provides an extended temperature range of -25°C to +80°C. The temperature is settable in 1°C increments and can maintain accuracy of + or – 0.1°C. Proven performance results are, improved long-term stability and precision, no dilution for volatile organic solvents, reduced oxide interferences (ICP-MS), enhanced sensitivity and accuracy, lower detection limits, faster washout, and reduced hazardous waste/disposal fees.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3182