Learn More
Glass Expansion IsoMist XS Ethernet Cable Replacement
SDP
Supplier:  Glass Expansion 708031990
Encompass_Preferred
IsoMist-XS Ethernet Cable. A replacement ethernet cable for Glass Expansion’s IsoMist-XS. Compatible with all IsoMist XS Kits. This IsoMist XS is programmable and temperature-controlled, with a glass cyclonic spray chamber now featuring an improved thermodynamic design that provides an extended temperature range of -25°C to +80°C. The temperature is settable in 1°C increments and can maintain accuracy of + or – 0.1°C. Proven performance results are, improved long-term stability and precision, no dilution for volatile organic solvents, reduced oxide interferences (ICP-MS), enhanced sensitivity and accuracy, lower detection limits, faster washout, and reduced hazardous waste/disposal fees.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3024