Learn More
Medchemexpress LLC HY-B0329 100mg Medchemexpress, Isoniazid CAS:54-85-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0329100MG
Encompass_Preferred
Medchemexpress, HY-B0329 100mg Isoniazid CAS:54-85-3 Isoniazid (INH) is a prodrug and must be activated by a bacterial catalase-peroxidase enzyme KatG. Isoniazid is bactericidal to rapidly dividing mycobacteria and has anti-tuberculostatic activity[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2473
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.