Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonicotinic Acid N-Oxide 98.0+%, TCI America™
SDP

Catalog No. I050025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I050025G 25 g
I0500100G 100 g
I0500250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I050025G Supplier TCI America Supplier No. I050025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13602-12-5 | C6H5NO3 | 139.11 g/mol

Chemical Identifiers

CAS 13602-12-5
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006209
InChI Key QCWTWMJMLSKQCJ-UHFFFAOYSA-N
Synonym isonicotinic acid n-oxide, 4-carboxypyridine 1-oxide, pyridine-4-carboxylic acid n-oxide, 4-pyridinecarboxylic acid 1-oxide, 4-pyridinecarboxylic acid, 1-oxide, pyridine-4-carboxylic acid 1-oxide, 4-carboxypyridin-1-ium-1-olate, isonicotinic acid 1-oxide, 1-hydroxypyridine-4-carboxylic acid, pyridin-n-oxide-4-carboxylic acid
PubChem CID 26130
IUPAC Name 4-carboxypyridin-1-ium-1-olate
SMILES OC(=O)C1=CC=[N+]([O-])C=C1

Specifications

Melting Point 270°C
Quantity 25 g
Formula Weight 139.11
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinic Acid N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.