Learn More
Selleck Chemical LLC Isonipecotic acid
SDP
Supplier:  Selleck Chemical LLC S561225MG
Encompass_Preferred
Isonipecotic acid (4-Piperidinecarboxylic acid 4-Carboxypiperidine Hexahydroisonicotinic acid) is a heterocyclic compound that mainly acts as a GABAA receptor partial agonist
Catalog No. 50-000-03574
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.