Learn More
Sigma Organic Chemistry Isopentyl 2-furoate | 1G | 615-12-3 | MFCD00059015
Supplier: Sigma Organic Chemistry CDS0005381G

Isopentyl 2-furoate , 1G
About this Item:
CAS #: 615-12-3
MDL #: MFCD00059015
Molecular Weight: 182.22
UNSPSC Code: 12352200
Chemical Formula: C10H14O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.