Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophthalaldehyde, 98%

Catalog No. AAA1578814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1578814 25 g
AAA1578806 5 g
AAA1578822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1578814 Supplier Thermo Scientific Chemicals Supplier No. A1578814
Add to Cart

CAS: 626-19-7 | C8H6O2 | 134.134 g/mol

Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-19-7
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00003372
InChI Key IZALUMVGBVKPJD-UHFFFAOYSA-N
Synonym isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x
PubChem CID 34777
IUPAC Name benzene-1,3-dicarbaldehyde
SMILES C1=CC(=CC(=C1)C=O)C=O

Specifications

Melting Point 85°C to 91°C
Boiling Point 136°C (17 mmHg)
Quantity 25 g
Beilstein 1561038
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 134.13
Percent Purity 98%
Chemical Name or Material Isophthalaldehyde
EINECSNumber 210-935-8
RTECSNumber NT1981000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.