Learn More
Medchemexpress LLC HY-N0769 5mg Medchemexpress, Isopimpinellin CAS:482-27-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07695MG
Encompass_Preferred
Medchemexpress, HY-N0769 5mg Isopimpinellin CAS:482-27-9 Isopimpinellin, an orally active compound isolated from the roots of Pimpinella saxifrage. Isopimpinellin blocks DNA adduct formation and skin tumor initiation by 7,12-dimethylbenz[a]anthracene. Isopimpinellin possesses anti-leishmania effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2821
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.