Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Isopore™ Membrane Filters, 0.4 μm

Catalog No. HTTP03700 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.4 μm
1.2 μm
Diameter (Metric):
13 mm
25 mm
37 mm
47 mm
90 mm
Color:
Black
White
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric) Color
HTTP03700 0.4 μm 37 mm White
HTTP01300 0.4 μm 13 mm White
RTTP01300 1.2 μm 13 mm White
HTTP02500 0.4 μm 25 mm White
HTBP02500 0.4 μm 25 mm Black
HTTP04700 0.4 μm 47 mm White
HTBP04700 0.4 μm 47 mm Black
HTTP09030 0.4 μm 90 mm White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. HTTP03700 Supplier MilliporeSigma™ Supplier No. HTTP03700
Add to Cart
Add to Cart

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

TRUSTED_SUSTAINABILITY

Specifications

Airflow Rate ≥7 L/min/psi
Bubble Point ≥1.5 bar
Color White
Filter Type Screen Filter
Material Polycarbonate
Pore Size 0.4 μm
Filter Code HTTP
For Use With (Application) For analysis of airborne contaminants & other particles using optical or electron microscopy
Porosity 10 to 20%
Quantity 100/Pk.
Surface Type Plain
Temperature Range (Metric) 140°C
Thickness (Metric) 28 μm
Type Filter Membranes
Diameter (Metric) 37 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.