Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Isopore™ Membrane Filters, 0.8 μm
GSA_VA

Catalog No. ATTP02500 Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (Metric):
13 mm
25 mm
47 mm
142 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
ATTP02500 25 mm
ATTP01300 13 mm
ATTP04700 47 mm
ATTP14250 142 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ATTP02500 Supplier MilliporeSigma™ Supplier No. ATTP02500
Add to Cart
Edge
Add to Cart

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features and Benefits:
  • Membrane structure retains particles on the surface, simplifying counting and analysis
  • Isopore™ membranes do not stain, resulting in low background interference
  • Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
  • Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
  • Applications: Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bubble Point 8.5 psi
Color White
Disposable Reusable
Filter Type Screen Filter
Format Disc
Material Polycarbonate
Pore Size 0.8 μm
Sterility Non-sterile
Flow Rate Avg. 85 mL/min. x cm2 (typical results (at 10 psi);125 mL/min. x cm2/psi)
For Use With (Application) Air Analysis, Cell Biology, Cell Culture, Bioassays, Cytology, Environmental Analysis, Epifluorescence
Gas Flow Rate 20.0 L/min./cm2
Hydrophobicity Hydrophilic
No. per Case 100 Pk.
Quantity 100 Pk.
Type Filter Membranes
Wettability Hydrophilic
Diameter (English) 1 in.
Diameter (Metric) 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.