Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoprene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018312 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8312 10 mL
11-102-0325 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8312 Supplier MilliporeSigma Supelco Supplier No. 5924010MLF
Add to Cart
Add to Cart

Isoprene is a reactive aliphatic hydrocarbon that originates as a byproduct of cholesterol synthesis.

TRUSTED_SUSTAINABILITY

Specifications

CAS 78-79-5
Solution Contains ˜0.02% 4-tert-butylcatechol as stabilizer
Vapor Pressure 8.82 psi (20°C)
Vapor Density 2.35 (vs. air)
Density 0.681 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH2=CHC(CH3)=CH2
MDL Number MFCD00008600
Melting Point -146°C
Boiling Point 34°C
Synonym 2-Methyl-1,3-butadiene
Quantity 10 mL
Formula Weight 68.12
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.