Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 70% (v/v), Ricca Chemical
SDP

Catalog No. R42100004C1 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R42100004C1 4 L Safety-coated Glass Bottle
4210-16 500 mL Natural Poly Bottle
4210-32 1 L Natural Poly Bottle
42101G 4 L Amber Glass Bottle
4210-1 4 L Natural Poly Bottle
42102-5 10 L Cubitainer
4210-5 20 L Cubitainer
42105HP 20 L Hard Pack
421055 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. R42100004C1 Supplier Ricca Chemical Company Supplier No. R42100004C1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) Mixture
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Min % 29.0, 69.0
CAS Max % 31.0, 71.0
Appearance Colorless Liquid
Quantity 4 L
Packaging Safety-coated Glass Bottle
Specific Gravity 0.874 to 0.882
Synonym isopropanol
Boiling Point 82°C
Percent Purity 68 to 72% v/v
Grade Laboratory
Chemical Name or Material Isopropanol
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.