Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, ACS Reagent (2-Propanol; Isopropyl Alcohol), Reagents

Catalog No. 01338118
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
Glass Bottle
ISOPROPANOL (IPA) ACS
Poly Bottle
Safety Coated Glass
Steel Pail
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-338-118 4 L Glass Bottle
01-338-124 10 L Steel Pail
01-338-123 1 L Glass Bottle
01-338-122 200 L ISOPROPANOL (IPA) ACS
01-338-121 20 L Steel Pail
01-338-120 20 L Cubitainer
01-338-119 4 L Poly Bottle
01-338-117 4 L Safety Coated Glass
01-338-116 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 01-338-118 Supplier Reagents Holdings Llc Supplier No. C11215004C
Add to Cart
Add to Cart

Isopropanol, ACS Reagent (2-Propanol; Isopropyl Alcohol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7808
Boiling Point 82.5°C
Grade ACS
Chemical Name or Material Isopropanol, ACS Reagent (2-Propanol, Isopropyl Alcohol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.