Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl 2-Methylbutyrate 98.0+%, TCI America™
SDP

Catalog No. I073625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I073625ML 25 mL
I0736500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I073625ML Supplier TCI America Supplier No. I073625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66576-71-4
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00085203
InChI Key DIRDKDDFAMNBNY-UHFFFAOYSA-N
Synonym 2-Methylbutyric Acid Isopropyl Ester
PubChem CID 522214
IUPAC Name propan-2-yl 2-methylbutanoate
SMILES CCC(C)C(=O)OC(C)C

Specifications

Color Colorless
Boiling Point 144°C
Quantity 25 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl 2-Methylbutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.