Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl 4-Hydroxybenzoate 99.0+%, TCI America™
SDP

Catalog No. I081725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I081725G 25 g
I08175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I081725G Supplier TCI America Supplier No. I081725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4191-73-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016468
InChI Key CMHMMKSPYOOVGI-UHFFFAOYSA-N
Synonym isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester
PubChem CID 20161
IUPAC Name propan-2-yl 4-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 86°C
Color White
Boiling Point 160°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥99.0% (T)
Physical Form Powder or Flakes
Chemical Name or Material Isopropyl 4-Hydroxybenzoate [for Biochemical Research]
EINECSNumber (3)-1585
RTECSNumber DH2250000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.