Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl acetate, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018755 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8755 10 mL
11-101-9332 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8755 Supplier MilliporeSigma Supelco Supplier No. 9087110MLF
Add to Cart
Add to Cart
Isopropyl acetate is an isopropyl ester of acetic acid. It participates in the mesoporous Al-MCM-41 (Si/Al = 55 and 104) and Al, Zn-MCM-41 (Si/(Al+Zn) = 52) molecular sieves catalyzed alkylation of m-cresol. It is widely used for the incorporation of aroma to various cosmetics and food products. Vapor-liquid equilibria for its binary mixture with CO2 at higher pressures has been evaluated. It is a colorless, flammable liquid, having a pleasant fruity type of odor.
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-21-4
Vapor Pressure 47 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.872 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3COOCH(CH3)2
MDL Number MFCD00008877
Melting Point -73°C
Boiling Point 85°C to 91°C
Synonym Acetic acid isopropyl ester
Quantity 10 mL
Formula Weight 102.13
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.