Learn More
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Specifications
| Melting Point | -73°C |
| Color | Colorless |
| Density | 0.872g/cm³ |
| Boiling Point | 88.8°C |
| Assay Percent Range | ≥99.0 % |
| Packaging | Glass Bottle |
| Linear Formula | CH3COOCH(CH3)2 |
| Quantity | 2.5 L |
| Vapor Pressure | 61mbar at 20°C |
| Formula Weight | 102.13 |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.