Learn More
Isopropyl Alcohol, 99%, EP, BP, JP, USP, 99%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, USP, EP, BP, JP has many uses, including as a pharmaceutical raw material for preparing USP Rubbing Alcohol for relief of muscle stiffness and an infection preventative for minor scrapes and burns. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Appearance | Clear |
| Quantity | 4 L |
| Packaging | Amber Glass Bottle |
| Refractive Index | 1.376-1.378 |
| Specific Gravity | 0.785-0.788 |
| Water | 0.5% |
| UN Number | UN1219 |
| Boiling Point | 78.3°C |
| Percent Purity | 99% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.