Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 99%, EP, BP, JP, USP, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603531 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-531 20 L Steel Can
18-603-525 500 mL Amber Glass Bottle
18-603-532 1 L Amber Glass Bottle
18-603-526 6 x 500 mL Amber Glass Bottle
18-603-527 4 L Amber Glass Bottle
18-603-528 4 L Poly Bottle
18-603-533 6 x 1 L Amber Glass Bottle
18-603-529 4 x 4 L Amber Glass Bottle
18-603-530 4 x 4 L Poly Bottle
18-603-534 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18-603-531 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IS11520LTMT
Add to Cart
Add to Cart

IS115, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, USP, EP, BP, JP has many uses, including as a pharmaceutical raw material for preparing USP Rubbing Alcohol for relief of muscle stiffness and an infection preventative for minor scrapes and burns. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100%
Appearance Clear
Quantity 20 L
Packaging Steel Can
Refractive Index 1.376-1.378
Specific Gravity 0.785-0.788
Water 0.5%
UN Number UN1219
Boiling Point 78.3°C
Percent Purity 99%
Grade Multi-Compendial
Chemical Name or Material Isopropyl Alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.