Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Solstice

Catalog No. 6000154 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 gal.
19 L
4 x 4 L
6 x 1 L
204 L
Packaging:
Fiberboard Box
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-001-54 4 x 4 L Poly Bottle
60-001-56 6 x 1 L Poly Bottle
60-001-53 4 x 4 L Glass Bottle
60-001-55 5 gal. Fiberboard Box
60-001-51 19 L Metal Can
60-001-52 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-001-54 Supplier Solstice Supplier No. LP3234P
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100.0000%
Density 0.785 g/cm3
Quantity 4 x 4 L
Flash Point 12°C
Packaging Poly Bottle
UN Number UN1219
Water Passes test
Beilstein 635639
Boiling Point 82.3°C
Solubility Information Miscible in Water at 20°C
Formula Weight 60.11
Percent Purity >99.5%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Isopropyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.