Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, FCC, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603523 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-523 20 L Steel Can
18-603-515 500 mL Amber Glass Bottle
18-603-517 1 L Amber Glass Bottle
18-603-516 6 x 500 mL Amber Glass Bottle
18-603-518 4 L Amber Glass Bottle
18-603-519 4 L Poly Bottle
18-603-520 6 x 1 L Amber Glass Bottle
18-603-521 4 x 4 L Amber Glass Bottle
18-603-522 4 x 4 L Poly Bottle
18-603-524 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18-603-523 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I105620LT
Add to Cart
Add to Cart

I1056, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, FCC is used to sanitize food processing equipment. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100%
Acidity 10 mg/kg
Quantity 20 L
Packaging Steel Can
Specific Gravity 0.784
Water 0.2%
UN Number UN1219
Boiling Point 78.3°C
Percent Purity 99.7%
Grade FCC
Chemical Name or Material Isopropyl Alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.