Learn More
Isopropyl Alcohol, Reagent, ACS, 99.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Isopropyl Alcohol, Reagent, ACS is a compound which is colorless, flammable and has a very strong odor. Isopropyl alcohol has many uses, including as an analytical solvent reagent in the laboratory. Other chemical applications include use as a chemical synthesis intermediate, disinfectant, fuel additive and as a biological specimen preservative.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 4 x 4 L |
| Packaging | Poly Bottle |
| Residue after Evaporation | 0.001% |
| Water | 0.2% |
| UN Number | UN1219 |
| Boiling Point | 78.3°C |
| Percent Purity | 99.5% |
| Grade | Reagent |
| Chemical Name or Material | Isopropyl Alcohol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.