Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Isopropyl Alcohol Spray

Catalog No. 19090659 Shop All Medique Products
Change view
Click to view available options
Volume (English):
2 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-090-659 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-659 Supplier Medique Supplier No. 26802
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Topical skin cleaner containing active ingredient Isopropyl Alcohol 70%.

  • Contains active ingredient isopropyl alcohol 70%

Specifications

Active Ingredients Isopropyl Alcohol 70%
Product Type Isopropyl Alcohol Spray
For Use With (Application) Topical skin cleaner
Volume (English) 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.