Learn More
Sigma Organic Chemistry Isopropyl benzoate | 1G | 939-48-0 | MFCD00048289
Supplier: Sigma Organic Chemistry CDS0005761G

Isopropyl benzoate , 1G
About this Item:
CAS #: 939-48-0
MDL #: MFCD00048289
Molecular Weight: 164.2
UNSPSC Code: 12352200
Chemical Formula: C10H12O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.