Learn More
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Shop All Thermo Scientific ProductsDescription
- Induces the lac operon in E. coli by binding and inactivating the lac repressor.
- Selection for lac Y mutants.
- Selection for recombinant plasmids containing β-galactosidase fusion proteins.
- Induces the cellular content of lactose permease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Induces lac operon in E. coli by binding and inactivating lac repressor
- Selection for lac Y mutants
- Selection for recombinant plasmids containing beta-galactosidase fusion proteins
- Induces cellular content of lactose permea
Specifications
Specifications
| Melting Point | 109°C to 115°C |
| Packaging | Plastic bottle |
| Quantity | 5 g |
| Beilstein | 4631 |
| Formula Weight | 238.3 |
| Percent Purity | ≥99% |
| Grade | Ultrapure |
| Physical Form | Powder |
| Chemical Name or Material | Isopropyl-beta-D-thiogalactopyranoside (IPTG) |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 206-703-0 |
| TSCA | Yes |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.