accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CS(S)O(R)OH(R)OH(S)HO(R)HO

CAS RN 367-93-1

IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
Synonyms: IPTG isopropyl-β-D-thiogalactoside isopropyl β-D-thiogalactoside isopropyl thiogalactoside
Molecular Weight (g/mol): 238.3
Molecular Formula: C9H18O5S
InChi Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N
SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (4)
  • (33)
Quantity 
  • (11)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (14)
  • (9)
  • (4)
  • (1)
Grade 
  • (9)
  • (17)
  • (4)
Percent Purity 
  • (2)
  • (13)
  • (1)
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (16)
  • (9)
  • (1)
  • (5)
  • (1)
  • (11)
  • (3)
  • (9)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (33)
  • (11)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (14)
  • (9)
  • (4)
  • (1)
  • (9)
  • (17)
  • (4)
  • (2)
  • (13)
  • (1)
  • (1)
  • (6)
  • (3)
  • (1)