Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl bromoacetate, 97%

Catalog No. AC412810250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412810250 25 g Glass bottle
AC412811000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412810250 Supplier Thermo Scientific Chemicals Supplier No. 412810250
Add to Cart

CAS: 29921-57-1 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29921-57-1
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00009886
InChI Key JCWLEWKPXYZHGQ-UHFFFAOYSA-N
Synonym isopropyl bromoacetate, acetic acid, bromo-, 1-methylethyl ester, acetic acid, 2-bromo-, 1-methylethyl ester, isopropyl 2-bromoacetate, brch2c o och ch3 2, bromoacetic acid isopropyl ester, i-propyl bromoacetate, acmc-1ciu8, isopropyl .alpha.-bromoacetate
PubChem CID 121593
IUPAC Name propan-2-yl 2-bromoacetate
SMILES CC(C)OC(=O)CBr

Specifications

Color Colorless to Yellow
Density 1.3990g/mL
Boiling Point 59.0°C to 61.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460
Linear Formula BrCH2CO2CH(CH3)2
Quantity 25 g
Specific Gravity 1.399
Formula Weight 181.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isopropyl bromoacetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 249-956-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.