Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., Dioxane Crystalline), Fisher BioReagents™
GSA_VA

Catalog No. BP162010 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1620-10 10 g Glass Bottle
BP1620-1 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1620-10
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 122°C
Color White
Quantity 10 g
Packaging Glass Bottle
Physical Form Solid
Merck Index 15, 5126
Formula Weight 238.3
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May form explosive peroxides. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:2
Recommended Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.