Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl-β-D-Thiogalactopyranoside, MP Biomedicals™

Catalog No. MP21021012 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21021012 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21021012 Supplier MP Biomedicals, Inc Supplier No. 02102101.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Commonly used as a reagent in cloning procedures. Isopropyl-β-D-Thiogalactopyranoside, MP Biomedicals (IPTG) is an analogue of allolactose that binds specifically to the repressor protein of the lac operon and induces expression of β-galactosida

IPTG is a commonly used reagent in cloning procedures that require induction of b-galactosidase activity and is used in conjunction with X-Gal in blue-white color selection of recombinant bacterial colonies.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 105°C to 115°C
pH 5.0 to 7.0
Quantity 250 mg
Solubility Information Soluble in methanol and water (50mg/mL, clear, colorless solution).
Optical Rotation −26° to −33° (c=1, water)
Formula Weight 238.3
Percent Purity ≥99%
Grade Not Available
Concentration or Composition (by Analyte or Components) Working Concentration: The final concentration typically used in indicator plates is 0.2 mM.
Physical Form Powder
Recommended Storage 0°C, desiccate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.