Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Ether, 99.0% min., ACS, MilliporeSigma™

Catalog No. MPX189020 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX189020 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX189020 Supplier MilliporeSigma Supplier No. PX189020

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Color Colorless
Density 0.72g/cm3 (20°C)
Explosion Limit 1.0 to 21%(V)
Ignition Point 405°C
Assay Percent Range 99.0% min.
Quantity 200 L
Vapor Pressure 175 hPa (20°C)
Formula Weight 12.18
Physical Form Liquid
Chemical Name or Material Isopropyl Ether, Stabilized With 2, 6-Di-Tert-Butyl-4-Methylphenol (BHT), For Synthesis
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.