Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. M028025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M028025ML 25 mL
M0280500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M028025ML Supplier TCI America Supplier No. M028025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4655-34-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00048290
InChI Key BOQSSGDQNWEFSX-UHFFFAOYSA-N
Synonym Methacrylic Acid Isopropyl Ester
PubChem CID 20769
IUPAC Name propan-2-yl 2-methylprop-2-enoate
SMILES CC(C)OC(=O)C(=C)C

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 mL
UN Number 1992
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1039
RTECSNumber OZ5020000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.