Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Methyl Sulfone 97.0+%, TCI America™
SDP

Catalog No. I048425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I048425G 25 g
I04845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I048425G Supplier TCI America Supplier No. I048425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4853-74-1
Molecular Formula C4H10O2S
Molecular Weight (g/mol) 122.182
MDL Number MFCD00050415
InChI Key VTWYQAQIXXAXOR-UHFFFAOYSA-N
PubChem CID 138372
IUPAC Name 2-methylsulfonylpropane
SMILES CC(C)S(=O)(=O)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 122.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Methyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.