Learn More
Teknova Isopropyl Myristate Solution. 500mL, Sterile. 4 Pack.
SDP
Supplier:  Teknova I12004PK
Encompass
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a galactose analog that can be used to induce expression of cloned genes under control of the lac operon. It is often used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies.
Catalog No. 50-238-7375
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.