Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl N-phenylcarbamate, 96%, Thermo Scientific™

Catalog No. AAB2499814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2499814 25 g
AAB2499822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2499814 Supplier Thermo Scientific Chemicals Supplier No. B2499814

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-42-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00026382
InChI Key VXPLXMJHHKHSOA-UHFFFAOYSA-N
Synonym propham, isopropyl n-phenylcarbamate, isopropyl phenylcarbamate, triherbide, agermin, collavin, prophame, tuberit, tuberite, birgin
PubChem CID 24685
ChEBI CHEBI:82035
IUPAC Name propan-2-yl N-phenylcarbamate
SMILES CC(C)OC(=O)NC1=CC=CC=C1

Specifications

Melting Point 86°C to 89°C
Quantity 25 g
Formula Weight 179.22
Percent Purity 96%
Chemical Name or Material Isopropyl N-phenylcarbamate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-542-0
RTECSNumber FD9100000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.