Learn More
Sigma Organic Chemistry Isopropyl salicylate | 1G | 607-85-2 | MFCD00035703
Supplier: Sigma Organic Chemistry CDS0012741G

Isopropyl salicylate , 1G
About this Item:
CAS #: 607-85-2
MDL #: MFCD00035703
Molecular Weight: 180.2
UNSPSC Code: 12352200
Chemical Formula: C10H12O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.