Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylamine, 99%

Catalog No. p-4510095
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148920250 25 mL Glass bottle
AC148920010 1 L Glass bottle
AC148920025 2.5 L Glass bottle
AC148920100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC148920250 Supplier Thermo Scientific Chemicals Supplier No. 148920250
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 75-31-0 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-31-0
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
InChI Key JJWLVOIRVHMVIS-UHFFFAOYSA-N
Synonym isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
PubChem CID 6363
ChEBI CHEBI:15739
IUPAC Name propan-2-amine
SMILES CC(C)N

Specifications

Melting Point -101.0°C
Color White
Density 0.6900g/mL
Boiling Point 33.0°C to 34.0°C
Flash Point −37°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3725 to 1.3755
Linear Formula (CH3)2CHNH2
Quantity 25 mL
Beilstein 04,152
Fieser 13,164
Merck Index 15,5255
Specific Gravity 0.69
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether
Viscosity 0.3 mPa.s (20°C)
Formula Weight 59.11
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Isopropylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
EINECSNumber 200-860-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.