Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. I0166100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0166100G 100 g
I016625G 25 g
I0166500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0166100G Supplier TCI America Supplier No. I0166100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15572-56-2
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00050705
InChI Key ISYORFGKSZLPNW-UHFFFAOYSA-N
Synonym 2-Aminopropane Hydrochloride, Isopropylammonium Chloride
PubChem CID 6432137
IUPAC Name propan-2-amine;hydrochloride
SMILES CC(C)N.Cl

Specifications

Melting Point 160°C
Color White
Quantity 100 g
Formula Weight 95.57
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropylamine Hydrochloride
EINECSNumber (2)-0131
RTECSNumber NT9530000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.