Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylamine Hydroiodide 97.0+%, TCI America™
SDP

Catalog No. I09341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09341G 1 g
I09345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09341G Supplier TCI America Supplier No. I09341G

Chemical Identifiers

CAS 66735-20-4
Molecular Formula C3H10IN
Molecular Weight (g/mol) 187.024
InChI Key VMLAEGAAHIIWJX-UHFFFAOYSA-N
Synonym Isopropylammonium Iodide
PubChem CID 91972165
IUPAC Name propan-2-amine;hydroiodide
SMILES CC(C)N.I

Specifications

Color White
Quantity 1 g
Formula Weight 187.02
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropylamine Hydroiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.