Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylidenemalononitrile 98.0+%, TCI America™
SDP

Catalog No. I048825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I048825G 25 g
I0488500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I048825G Supplier TCI America Supplier No. I048825G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13166-10-4
Molecular Formula C6H6N2
Molecular Weight (g/mol) 106.128
MDL Number MFCD00082572
InChI Key NVBHWAQBDJEGEO-UHFFFAOYSA-N
Synonym 1,1-Dicyano-2-methyl-1-propene
PubChem CID 533473
IUPAC Name 2-propan-2-ylidenepropanedinitrile
SMILES CC(=C(C#N)C#N)C

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 g
UN Number 3276
Formula Weight 106.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropylidenemalononitrile
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.