Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline-5-sulfonic acid, 95%, Thermo Scientific™

Catalog No. AAH6173914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6173914 25 g
AAH6173906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6173914 Supplier Thermo Scientific Chemicals Supplier No. H6173914

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27655-40-9
Molecular Formula C9H7NO3S
Molecular Weight (g/mol) 209.219
MDL Number MFCD00134089
InChI Key YFMJTLUPSMCTOQ-UHFFFAOYSA-N
Synonym 5-isoquinolinesulfonic acid, 5-sulfoisoquinoline, 5-isoquinolinesulfonicacid, 5-sulphoisoquinoline, pubchem23533, acmc-1chvq, isoquinoline-5-sulphonic acid, ksc203s9f
PubChem CID 241599
IUPAC Name isoquinoline-5-sulfonic acid
SMILES C1=CC2=C(C=CN=C2)C(=C1)S(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
UN Number UN2585
Formula Weight 209.22
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Isoquinoline-5-sulfonic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.