Learn More
AdipoGen Isorhamnetin (5 mg)
SDP
Supplier:  AdipoGen CDXI0174M005
Encompass_Preferred
Chemical. CAS: 480-19-3. Formula: C16H12O7. MW: 316.26. Isorhamnetin is a natural flavonol aglycone that is the 3-methyl metabolite of quercetin. It shows diverse biological activities, including antioxidant, cardioprotective, anti-inflammatory, neuroprotective, antiobesity, antiviral, antituberculosis and anticancer properties. Isorhamnetin exerts anticancer activity, through induction of apoptosis and inhibition of cell proliferation and invasion. It suppresses colon cancer cell growth through the PI3K-Akt-mTOR pathway. It inhibits the NF-kappaB signaling pathway. Isorhamnetin selectively inhibited NLRP3 and AIM2 inflammasome activation and down-regulated expression of pro-inflammatory cytokines. It has powerful antioxidant action through its ability to scavenge free radicals and inhibits xanthine oxidase. Isorhamnetin is a PPARgamma antagonist and inhibits adipogenesis, with beneficial effect on the prevention and treatment of obesity.
Catalog No. 50-205-4512
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.